missing translation for 'onlineSavingsMsg'
Learn More
Learn More
beta II Tubulin B Antibody (OTI6D11), Alexa Fluor™ 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72505AF405
This item is not returnable.
View return policy
Description
beta II Tubulin B Monoclonal specifically detects beta II Tubulin B in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| beta II Tubulin B | |
| Monoclonal | |
| Alexa Fluor 405 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| TUBB2B | |
| Full length recombinant protein of human beta II Tubulin B (NP_821080) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat, Canine, Monkey | |
| IgG2a |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| OTI6D11 | |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| bA506K6.1, class II beta-tubulin isotype, DKFZp566F223, FLJ98847, MGC8685, tubulin beta-2B chain, tubulin, beta 2B, tubulin, beta polypeptide paralog | |
| Mouse | |
| Affinity Purified | |
| Signal Transduction | |
| 347733.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction