missing translation for 'onlineSavingsMsg'
Learn More
Learn More
beta II Tubulin B Antibody (OTI6D11) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72505
This item is not returnable.
View return policy
Description
beta II Tubulin B Monoclonal specifically detects beta II Tubulin B in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| beta II Tubulin B | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1:400-1:2000, Immunocytochemistry/Immunofluorescence 1:100 | |
| bA506K6.1, class II beta-tubulin isotype, DKFZp566F223, FLJ98847, MGC8685, tubulin beta-2B chain, tubulin, beta 2B, tubulin, beta polypeptide paralog | |
| Mouse | |
| Affinity Purified | |
| Signal Transduction | |
| 347733 | |
| Human, Mouse, Rat, Canine, Monkey | |
| Lyophilized |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| OTI6D11 | |
| Unconjugated | |
| Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative | |
| TUBB2B | |
| Full length human recombinant protein of human TUBB2B(NP_821080) produced in HEK293T cell. | |
| 100 μg | |
| Primary | |
| Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended. | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction