missing translation for 'onlineSavingsMsg'
Learn More
Learn More
beta-III Tubulin Antibody (2G10), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-37816
This item is not returnable.
View return policy
Description
beta-III Tubulin Monoclonal specifically detects beta-III Tubulin in Human, Mouse, Rat, Porcine, Bovine, Guinea Pig, Hamster, Rabbit samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.
Specifications
| beta-III Tubulin | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500 - 1:2000, Flow Cytometry 1:50, Immunohistochemistry 1:50 - 1:500, Immunocytochemistry/Immunofluorescence 1:50 - 1:200, Immunoprecipitation 5 ug | |
| beta-4, TUBB4 CFEOM3A, tubulin beta-3 chain, Tubulin beta-4 chain, Tubulin beta-III, tubulin, beta 3, TUJ1 antigen, TUJ-1 antigen | |
| Mouse | |
| 50 kDa | |
| 20 μL | |
| Cell Cycle and Replication, Cellular Markers, Cytoskeleton Markers, Neuroscience | |
| 10381 | |
| Human, Mouse, Rat, Pig, Bovine, Guinea Pig, Hamster, Rabbit | |
| Purified |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
| 2G10 | |
| Unconjugated | |
| PBS, 1.0 mg/ml BSA and 30% Glycerol with 0.05% Sodium Azide | |
| TUBB3 | |
| A synthetic peptide corresponding to amino acids 436-450 from rat neuronal specific beta-3 tubulin. | |
| Protein A purified | |
| RUO | |
| Primary | |
| Western blot analysis of MA1-118Detects an ∼50 kDa protein in neuronal-type cells. In bovine, a unknown band at ∼32 kDa is also detected. MA1-118 shows specificity to beta-3 Tubulin and is non-reactive to lysates from non-neuronal cell types (e.g. HeLa cell lysate). | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction