missing translation for 'onlineSavingsMsg'
Learn More
Learn More
beta-III Tubulin Antibody (rTUBB3/7406), Alexa Fluor™ 750, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-20907AF750
This item is not returnable.
View return policy
Description
beta-III Tubulin Monoclonal antibody specifically detects beta-III Tubulin in Human samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)
Specifications
| beta-III Tubulin | |
| Monoclonal | |
| Alexa Fluor 750 | |
| beta-4, TUBB4 CFEOM3A, tubulin beta-3 chain, Tubulin beta-4 chain, Tubulin beta-III, tubulin, beta 3, TUJ1 antigen, TUJ-1 antigen | |
| Recombinant fragment (around aa250-450) of human beta-III Tubulin (exact sequence is proprietary) | |
| 0.1 mL | |
| Cell Biology, Cell Cycle and Replication, Cellular Markers, Neuroscience, Phospho Specific | |
| 10381 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Western Blot, Immunohistochemistry (Paraffin) | |
| rTUBB3/7406 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction