missing translation for 'onlineSavingsMsg'
Learn More
Learn More
beta-III Tubulin Antibody (TUBB3/3732), DyLight 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08455DL594
This item is not returnable.
View return policy
Description
beta-III Tubulin Monoclonal antibody specifically detects beta-III Tubulin in Human, Canine samples. It is validated for Western Blot, Immunohistochemistry (Paraffin), Protein Array
Specifications
| beta-III Tubulin | |
| Monoclonal | |
| DyLight 594 | |
| beta-4, TUBB4 CFEOM3A, tubulin beta-3 chain, Tubulin beta-4 chain, Tubulin beta-III, tubulin, beta 3, TUJ1 antigen, TUJ-1 antigen | |
| A synthetic peptide (aa 437-450) of human Tubulin beta 3 protein (TUBB3); coupled to KLH. (Uniprot: Q13509) | |
| 100 μg | |
| Cell Biology, Cell Cycle and Replication, Cellular Markers, Neuroscience | |
| 10381 | |
| Store at 4°C in the dark. | |
| IgG2a κ |
| Western Blot, Immunohistochemistry (Paraffin), Protein Array | |
| TUBB3/3732 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Canine | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction