Learn More
beta-III Tubulin Antibody (TUBB3/7090R), Allophycocyanin, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP3-14268APC
Product Code. 18660157
  • € 690.00 / 0.10mL
Description
beta-III Tubulin Monoclonal antibody specifically detects beta-III Tubulin in Human samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
beta-III Tubulin
Monoclonal
APC
beta-4, TUBB4 CFEOM3A, tubulin beta-3 chain, Tubulin beta-4 chain, Tubulin beta-III, tubulin, beta 3, TUJ1 antigen, TUJ-1 antigen
A synthetic peptide (aa 437-450) of human Tubulin beta 3 protein (TUBB3); coupled to KLH. (Uniprot: Q13509 )
0.1 mL
Cell Biology, Cell Cycle and Replication, Cellular Markers, Neuroscience, Phospho Specific
10381
Store at 4°C in the dark.
IgG
Immunohistochemistry (Paraffin)
TUBB3/7090R
PBS
Rabbit
Protein A or G purified