Learn More
Invitrogen™ Beta III Tubulin Polyclonal Antibody
Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA5143785
Descripción
This antibody was cross-adsorbed to phospho-betaIII-Tubulin (Ser-444) peptide before affinity purification using unphosphorylated betaIII-Tubulin (Ser-444) peptide (without carrier). The antibody detects a 50 kDa* protein corresponding to the molecular mass of unphosphorylated betaIII-Tubulin on SDS-PAGE immunoblots of purified brain tubulin and mouse brain tissue.
The betaIII-tubulin isoform is present dominantly in cells of neuronal origin and it is one of the earliest markers of neuronal differentiation. It is regarded as a specific probe for the cells of neuronal origin as well as for the tumours originating from these cells. The betaIII-tubulin is most abundant in cells of neuronal origin, but was also detected in Sertoli cells of the testis and transiently in non-neuronal embryonic tissues.
Especificaciones
| Beta III Tubulin | |
| Polyclonal | |
| Unconjugated | |
| TUBB3 | |
| 3200002H15Rik; beta 3 tubulin; beta tubulin 3; beta-4; betaIII-tubulin; CDCBM; CDCBM1; CFEOM3; CFEOM3A; class III beta-tubulin; FEOM3; I79_000194; LOC100341581; M(beta)3; M(beta)6; mc1R; melanocortin receptor 1; Neuron specific beta III Tubulin; neuron-specific class III beta-tubulin; TUBB; Tubb3; TUBB4; tubulin beta; tubulin beta 3 class III; Tubulin beta-3; tubulin beta-3 chain; Tubulin beta-4; Tubulin beta-4 chain; tubulin beta-III; tubulin, beta 3 class III; tubulin, beta, 3; tubulin, beta, 4; Unknown (protein for MGC:134115) | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10381, 22152, 246118 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.125 mg/mL | |
| PBS with 1mg/mL BSA, 50% glycerol and 0.05% sodium azide | |
| Q13509, Q4QRB4, Q9ERD7 | |
| TUBB3 | |
| Unphosphorylated betaIII-Tubulin synthetic peptide (coupled to KLH) corresponding to amino acid residues around serine 444 of human betaIII-Tubulin. This sequence is not found in bI or betaII-Tubulin isotypes, but is well conserved in betaIII-Tubulins from rat and mouse. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.