Learn More
beta-NGF Antibody (MEDI-578) [Janelia Fluor™ 635], Novus Biologicals™
Human Monoclonal Antibody
Brand:  Novus Biologicals (Bio-Techne) NBP3-28861JF635
Product Code. 30171332
  • € 431.00 / 0.10mL
Description
beta-NGF Monoclonal antibody specifically detects beta-NGF in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY
Specifications
Specifications
beta-NGF
Monoclonal
Janelia Fluor 635
Beta nerve growth factor precursor, beta-nerve growth factor, Beta-NGF, HSAN5, MGC161426, nerve growth factor (beta polypeptide), nerve growth factor, beta subunit, NGFBMGC161428
NGF/ bNGF
0.1 mL
Cancer, Neuroscience
4803
Store at 4°C in the dark.
IgG4
Flow Cytometry, ELISA, Functional Assay
MEDI-578
50mM Sodium Borate
Human
Protein A purified