missing translation for 'onlineSavingsMsg'
Learn More
Learn More
beta-NGF Rabbit anti-Human, Clone: B64, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89766-100UL
This item is not returnable.
View return policy
Description
beta-NGF Monoclonal specifically detects beta-NGF in Human samples. It is validated for ELISA, Sandwich ELISA Capture.
Specifications
| beta-NGF | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| B64 | |
| ELISA 1:5000-1:10000, Sandwich ELISA Capture 1:250-1:2000 | |
| Beta nerve growth factor precursor, beta-nerve growth factor, Beta-NGF, HSAN5, MGC161426, nerve growth factor (beta polypeptide), nerve growth factor, beta subunit, NGFBMGC161428 | |
| Recombinant Human beta-NGF Protein | |
| 100 μg | |
| Cancer, Neuroscience | |
| 4803 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction