missing translation for 'onlineSavingsMsg'
Learn More
Learn More
beta Sarcoglycan Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-38648SS
This item is not returnable.
View return policy
Description
beta Sarcoglycan Polyclonal antibody specifically detects beta Sarcoglycan in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)
Specifications
| beta Sarcoglycan | |
| Polyclonal | |
| Unconjugated | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 1.0 mg/mL | |
| Immunohistochemistry 1:100 - 1:500, Immunohistochemistry-Paraffin 1:100 - 1:500 | |
| 43 kDa dystrophin-associated glycoprotein, 43DAG, A3bbeta-SG, beta-sarcoglycan, beta-sarcoglycan(43kD dystrophin-associated glycoprotein), Beta-SG, LGMD2E, limb girdle muscular dystrophy 2E (non-linked families), sarcoglycan, beta (43kD dystrophin-associated glycoprotein), sarcoglycan, beta (43kDa dystrophin-associated glycoprotein), SGC | |
| Partial recombinant protein made to the C-terminal portion of human beta Sarcoglycan [UniProt Q16585]. | |
| 0.025 mg | |
| Signal Transduction | |
| 6443 | |
| Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction