missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Beta sarcoglycan Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11936-1-AP-20UL
This item is not returnable.
View return policy
Description
The function of this protein remains unknown.Specifications
| Beta sarcoglycan | |
| Polyclonal | |
| Unconjugated | |
| SGCB | |
| 43DAG, A3b, Beta sarcoglycan, Beta SG, LGMD2E, SGC, SGCB | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 24051, 6443, 680229 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P82349, Q16585 | |
| SGCB | |
| Beta sarcoglycan Fusion Protein Ag2533 | |
| 20 μL | |
| Primary | |
| Mouse, Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction