Learn More
beta-Sitosterol, with ca. 10% campesterol, ca. 75% beta-sitosterol
Product Code. p-7203417 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
Unit Size:
100g
5g
Product Code. Quantity unitSize
10654992 5 g 5g
10664992 100 g 100g
2 options
Product Code. 10654992
Brand: Thermo Scientific Acros 132720050
Request Bulk or Custom Format
CAS: 83-46-5 | C29H50O | 414.72 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 83-46-5
Molecular Formula C29H50O
InChI Key KZJWDPNRJALLNS-VJSFXXLFSA-N
Synonym beta-sitosterol, sitosterol, cupreol, quebrachol, 22,23-dihydrostigmasterol, azuprostat, triastonal, cinchol, rhamnol, harzol
PubChem CID 222284
ChEBI CHEBI:27693
Specifications
Color White
Loss on Drying 3.5% max.
Infrared Spectrum Authentic
Quantity 5 g
Specific Rotation -24° to -32° (20°C, 589nm) (c=2, CHCl3) Typically
SMILES CCC(CCC(C)C1CCC2C1(CCC3C2CC=C4C3(CCC(C4)O)C)C)C(C)C
IUPAC Name (3S,8S,9S,10R,13R,14S,17R)-17-[(2R,5R)-5-ethyl-6-methylheptan-2-yl]-10,13-dimethyl-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-3-ol
Molecular Weight (g/mol) 414.718
Formula Weight 414.70
Percent Purity ≥75%
Physical Form Powder
Chemical Name or Material β-Sitosterol