missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ beta-Synuclein Antibody (14) [Alexa Fluor™ 594], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-26905AF594
This item is not returnable.
View return policy
Description
beta-Synuclein Monoclonal antibody specifically detects beta-Synuclein in Human samples. It is validated for ELISA
Specifications
| beta-Synuclein | |
| Monoclonal | |
| Alexa Fluor 594 | |
| beta-synuclein, synuclein, beta | |
| This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human beta-Synuclein (Uniprot#: Q16143; aa 1-134). | |
| 0.1 mL | |
| Alzheimers Research, Neuroscience | |
| 6620 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA | |
| 14 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction