missing translation for 'onlineSavingsMsg'
Learn More
Learn More
beta-Synuclein Antibody, HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-05529H
This item is not returnable.
View return policy
Description
beta-Synuclein Monoclonal antibody specifically detects beta-Synuclein in Human, Mouse, Rat, Porcine, Bovine samples. It is validated for Western Blot, Immunohistochemistry
Specifications
| beta-Synuclein | |
| Monoclonal | |
| Western Blot, Immunohistochemistry | |
| beta-synuclein, synuclein, beta | |
| C-terminal peptide of human beta-Synuclein EPEGESYEDPPQEEYQEYEPEA coupled to KLH | |
| 0.1 mL | |
| Alzheimers Research, Neuroscience | |
| 6620 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| HRP | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Pig, Bovine | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction