missing translation for 'onlineSavingsMsg'
Learn More
Learn More
beta-Synuclein Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 357.00 - € 517.00
Specifications
| Antigen | beta-Synuclein |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:1000-2000, Immunocytochemistry/ Immunofluorescence 1:1000-2000 |
| Applications | Western Blot, Immunofluorescence |
| Classification | Polyclonal |
| Produktcode | Marke | Quantity | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Quantity | Preis | Menge & Verfügbarkeit | |||||
|
18662598
|
Novus Biologicals
NBP3-05556-100ul |
100 μg |
€ 517.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18679968
|
Novus Biologicals
NBP3-05556-50ul |
50 μg |
€ 357.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Beschreibung
beta-Synuclein Polyclonal antibody specifically detects beta-Synuclein in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ ImmunofluorescenceSpezifikation
| beta-Synuclein | |
| Western Blot 1:1000-2000, Immunocytochemistry/ Immunofluorescence 1:1000-2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| beta-synuclein, synuclein, beta | |
| C-terminal peptide of human beta-Synuclein EPEGESYEDPPQEEYQEYEPEA coupled to KLH | |
| Primary | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. |
| 1 mg/mL | |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Alzheimers Research, Neuroscience | |
| 50% PBS, 50% glycerol | |
| 6620 | |
| IgG1 | |
| Immunogen affinity purified |
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts