missing translation for 'onlineSavingsMsg'
Learn More
Learn More
beta Tubulin Antibody (BT7R), Janelia Fluor™ 549, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-37830JF549
This item is not returnable.
View return policy
Description
beta Tubulin Monoclonal antibody specifically detects beta Tubulin in Human, Mouse, Rat, Avian, Chicken, Primate, Rabbit samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Flow (Intracellular), CyTOF
Specifications
| beta Tubulin | |
| Monoclonal | |
| Janelia Fluor 549 | |
| beta polypeptide, MGC117247, Tubb5, tubulin beta chain, tubulin beta polypeptide, tubulin beta-1 chain, Tubulin beta-5 chain, tubulin, beta | |
| KLH conjugated peptide of the N-terminus of Beta-Tubulin. | |
| 0.1 mL | |
| Apoptosis, Breast Cancer, Cell Biology, Cellular Markers, Signal Transduction | |
| 203068 | |
| Store at 4°C in the dark. | |
| IgG2a |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence | |
| BT7R | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Avian, Chicken | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction