Learn More
Beta Tubulin Mouse anti-Human, CoraLite 488, Clone: 1D4A4, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL488-66240-100UL
Description
The Beta Tubulin antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human Beta Tubulin. This antibody recognizes human, mouse, rat, nematode, pig, zebrafish antigen. The Beta Tubulin antibody has been validated for the following applications: IF, WB analysis. This antibody is coralite™488.
Specifications
| Beta Tubulin | |
| Monoclonal | |
| 2000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Mouse, Rat, Nematode, Pig |
| Western Blot, Immunofluorescence | |
| 1D4A4 | |
| CoraLite 488 | |
| 100 μL | |
| 10381 | |
| IgG2a |