Learn More
Beta Tubulin Mouse anti-Human, CoraLite 488, Clone: 1D4A4, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech CL488-66240-100UL
Product Code. 16840756
  • € 189.00 / 100µL
Description
The Beta Tubulin antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human Beta Tubulin. This antibody recognizes human, mouse, rat, nematode, pig, zebrafish antigen. The Beta Tubulin antibody has been validated for the following applications: IF, WB analysis. This antibody is coralite™488.
Specifications
Specifications
Beta Tubulin
Monoclonal
2000 μg/mL
Mouse
Primary
Human, Mouse, Rat, Nematode, Pig
Western Blot, Immunofluorescence
1D4A4
CoraLite 488
100 μL
10381