missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Betacellulin/BTC Antibody (RM0154-9H1), DyLight 650, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-12053C
This item is not returnable.
View return policy
Description
Betacellulin/BTC Monoclonal specifically detects Betacellulin/BTC in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Betacellulin/BTC | |
| Monoclonal | |
| DyLight 650 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| BTC | |
| Mouse recombinant Betacellulin/BTC | |
| 0.1 mL | |
| Primary | |
| Mouse | |
| Purified |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| RM0154-9H1 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| betacellulin, probetacellulin | |
| Rat | |
| Protein A or G purified | |
| Biologically Active Proteins, Cell Cycle and Replication, Signal Transduction | |
| 685 | |
| Store at 4C in the dark. | |
| IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction