missing translation for 'onlineSavingsMsg'
Learn More

Betamipron, MedChemExpress

Product Code. 16498626
Encompass_Preferred
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
This item is not returnable. View return policy

Product Code. 16498626

Brand: Medchem Express HYB1127

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Betamipron is a chemical compound which is used together with Panipenem to inhibit Panipenem uptake into the renal tubule and prevent nephrotoxicity.

Betamipron is a chemical compound which is used together with Panipenem to inhibit Panipenem uptake into the renal tubule and prevent nephrotoxicity.

Specifications

Chemical Name or Material Betamipron
Physical Form Solid
CAS 3440-28-6
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C10H11NO3
Synonym N-Benzoyl-β-alanine
Solubility Information DMSO : 100 mg/mL (517.60 mM; Need ultrasonic)
SMILES O=C(O)CCNC(C1=CC=CC=C1)=O
Molecular Weight (g/mol) 193.2
Formula Weight 193.2
Percent Purity 99.66%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.