missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Betula pendula BETVIA Chimeric Recombinant Human Monoclonal Antibody (M0418)
GREENER_CHOICE

Product Code. 30089779
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30089779 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30089779 Supplier Invitrogen™ Supplier No. MA551583

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Human Recombinant Monoclonal Antibody

This antibody binds to Bet v 1, in particular to the naturally occurring isoforms Bet v 1.0101 and Bet v 1.0102, but not to Bet v 1.0112.

May be a general steroid carrier protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Betula pendula BETVIA Chimeric
Applications ELISA
Classification Recombinant Monoclonal
Clone M0418
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% ProClin 300
Gene Alias allergen Bet v 1-A; Allergen Bet v I-A; BETVI; Major pollen allergen Bet v 1-A
Host Species Human
Immunogen Bet v 1-A, a major allergen of the European white birch.
Purification Method Affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Plant
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgE λ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.