Learn More
Bevacizumab Mouse, Unconjugated, Monoclonal Antibody, AcroBiosystems
Product Code. 16427229
Change view
Click to view available options
Quantity:
1 mg
100 μg
Unit Size:
100µg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16427229 100 μg 100µg
16437229 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16427229 Supplier ACROBiosystems Supplier No. BEBY9100ug
Mouse Monoclonal Antibody
Bevacizumab Monoclonal antibody specifically detects Bevacizumab, recommended for ADA assay.
Specifications
Antigen Bevacizumab
Classification Monoclonal
Conjugate Unconjugated
Host Species Hybridoma
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Content And Storage -20°C