missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BEX4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA565688
This item is not returnable.
View return policy
Description
Immunogen sequence: ESKEELAANNL NGENAQQENE GGEQAPTQNE EESRHLGGGE GQKPGGNIRR GRVRRLVPN Highest antigen sequence identity to the following orthologs - mouse 47%, rat 42%.
Bex4 may play a role in microtubule deacetylation by negatively regulating the SIRT2 deacetylase activity toward alpha-tubulin and thereby participate to the control of cell cycle progression and genomic stability.
Specifications
| BEX4 | |
| Polyclonal | |
| Unconjugated | |
| Bex4 | |
| BEX family member 4; BEX1-like 1; BEX1-like protein 1; Bex4; BEXL1; brain expressed gene 4; brain expressed X-linked 4; brain expressed X-linked-like 1; brain expressed, X-linked 4; Brain-expressed X-linked protein 4; brain-expressed X-linked protein 4 homolog; FLJ10097; NADE3; Nerve growth factor receptor-associated protein 3; protein BEX4 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 56271 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9NWD9 | |
| Bex4 | |
| Recombinant Human BEX4. Recombinant protein control fragment (Product #RP-106362). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction