missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BFAR, Mouse anti-Human, Clone: 1C6, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00051283-M01.50ug
This item is not returnable.
View return policy
Description
Sequence: LRFEDIQQNNDIVQSLAAFQKYGNDQIPLAPNTGRANQQMGGGFFSGVLTALTGVAVVLLVYHWSSRESEHDLLVHKAVAKWTAEEVVLWLEQLGPWASLSpecifications
| BFAR | |
| Monoclonal | |
| Unconjugated | |
| bifunctional apoptosis regulator | |
| BAR/RNF47 | |
| Mouse | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Immunohistochemistry (Paraffin), KnockDown, Western Blot | |
| 1C6 | |
| In 1x PBS, pH 7.4 | |
| BC003054 | |
| BFAR | |
| BFAR (AAH03054, 101 a.a. ∼ 200 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 51283 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction