missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BFAR Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA572900
This item is not returnable.
View return policy
Description
BFAR Polyclonal Antibody for Western Blot, IHC (P), IP
The bifunctional apoptosis inhibitor (BFAR) is scaffold protein that integrates signaling components of the cells apoptosis-regulatory machinery. BFAR is a multidomain protein capable of inhibiting apoptosis induced by TNF-family death receptors ('extrinsic pathway') as well as mitochondria-dependent apoptosis ('intrinsic pathway'). Interaction of BFAR with Bcl-2 or Bcl-XL via a SAM domain may contribute to the anti-apoptotic properties of BFAR. In addition, BFAR contains a DED-like domain that is capable of suppressing apoptosis mediated at the receptor level. BFAR is also thought to be involved in the regulation of neuronal survival.
Specifications
| BFAR | |
| Polyclonal | |
| Unconjugated | |
| BFAR | |
| 3010001A07Rik; AI666707; AW107665; BAR; Bfar; bifunctional apoptosis inhibitor; bifunctional apoptosis regulator; RING finger protein 47; RNF47 | |
| Rabbit | |
| 50 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| Conc. Not Determined | |
| Whole serum with 0.05% sodium azide | |
| Q9NZS9 | |
| BFAR | |
| A recombinant protein corresponding to amino acids 1-139 of human BAR. | |
| RUO | |
| 51283 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction