missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BFSP1 Monoclonal Antibody (OTI1F1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA804040S
This item is not returnable.
View return policy
Description
BFSP1 Monoclonal antibody specifically detects BFSP1 in Human samples. It is validated for Western BlotSpecifications
| BFSP1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P03372 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 2099 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2a |
| Western Blot | |
| OTI1F1 | |
| Unconjugated | |
| ESR1 | |
| Beaded filament structural protein 1; beaded filament structural protein 1, filensin; BFSP1; CP115; CP94; CTRCT33; cytoskeletal protein, 115 KD; filensin; Lens fiber cell beaded-filament structural protein CP 115; lens intermediate filament-like heavy; LIFL-H; LOW QUALITY PROTEIN: filensin; RP4-531H16.2 | |
| Human recombinant protein fragment corresponding to amino acids 1-310 of human ESR1 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction