missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BFSP1 Monoclonal Antibody (OTI1F1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA804040
This item is not returnable.
View return policy
Description
More than 99% of the vertebrate ocular lens is comprised of terminally differentiated lens fiber cells. Two lens-specific intermediate filament-like proteins, CP49 (also known as phakinin) and the protein product of this gene, filensin, are expressed only after fiber cell differentiation has begun. Both proteins are found in a structurally unique cytoskeletal element that is referred to as the beaded filament (BF). Mutations in this gene are the cause of autosomal recessive cortical juvenile-onset cataract. Multiple transcript variants encoding different isoforms have been found for this gene.Specifications
| BFSP1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q12934 | |
| BFSP1 | |
| Human recombit protein fragment corresponding to amino acids 405-665 of human BFSP1 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Western Blot | |
| OTI1F1 | |
| Unconjugated | |
| BFSP1 | |
| CP115, CP94, CTRCT33, LIFL-H | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 631 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction