Learn More
BGJ 398, TRC
Specifications
Specifications
| Chemical Name or Material | 3-(2,6-Dichloro-3,5-dimethoxyphenyl)-1-[6-[(4-(4-ethylpiperazin-1-yl)phenyl)amino]pyrimidin-4-yl]-1-methylurea |
| CAS | 872511-34-7 |
| Molecular Formula | C26H31Cl2N7O3 |
| Quantity | 50 mg |
| Synonym | BGJ 398, Infigratinib, NVP-BGJ 398 |
| InChI Formula | InChI=1S/C26H31Cl2N7O3/c1-5-34-10-12-35(13-11-34)18-8-6-17(7-9-18)31-21-15-22(30-16-29-21)33(2)26(36)32-25-23(27)19(37-3)14-20(38-4)24(25)28/h6-9,14-16H,5,10-13H2,1-4H3,(H,32,36)(H,29,30,31) |
| SMILES | CCN1CCN(CC1)c2ccc(Nc3cc(ncn3)N(C)C(=O)Nc4c(Cl)c(OC)cc(OC)c4Cl)cc2 |
| IUPAC Name | 3-(2,6-dichloro-3,5-dimethoxyphenyl)-1-[6-[4-(4-ethylpiperazin-1-yl)anilino]pyrimidin-4-yl]-1-methylurea |
| Molecular Weight (g/mol) | 560.48 |