missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ BHLHB5 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA524711

Product Code. 13291679

  • € 578.00 / 400µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This antibody is predicted to react with chicken and mouse based on sequence homology.

BHLHB5 inhibits DNA binding of TCF3/E47 homodimers and TCF3 (E47)/NEUROD1 heterodimers and acts as a strong repressor of Neurod1 and Myod-responsive genes, probably by heterodimerization with class a basic helix-loop-helix factors. BHLHB5 despite the presence of an intact basic domain, does not bind to DNA.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

BHLHB5
Polyclonal
PBS with 0.09% sodium azide; pH 7.4
Q8NFJ8
BHLHE22
KLH conjugated synthetic peptide between 236-264 amino acids from the central region of human BHLHB5.
400 μL
Primary
Human
Antibody
IgG
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
BHLHE22
basic helix-loop-helix (bHLH) gene, class B, beta3; basic helix-loop-helix domain containing, class B, 5; basic helix-loop-helix domain containing, class B5; basic helix-loop-helix family member e22; basic helix-loop-helix family, member e22; Beta3; BHLHB5; bHLHe22; CAGL85; Class B basic helix-loop-helix protein 5; class E basic helix-loop-helix protein 22; Protein BETA3; TNRC20; trinucleotide repeat containing 20; trinucleotide repeat-containing gene 20 protein
Rabbit
Antigen affinity chromatography, Protein A
RUO
27319
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.