missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ BHLHE40 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16330005
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16330005 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16330005 Supplier Invitrogen™ Supplier No. PA583044

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: GRRFTMGERI GAIKQESEEP PTKKNRMQLS DDEGHFTSSD LISSPFLGPH PHQPPFCLPF YLIPPSATAY LPMLEKCWYP TSVPVLYPGL NASAAALSSF MNPDKISAPL LMPQRLPSPL PAHPSVDSSV LLQ Highest antigen sequence indentity to the following orthologs: Mouse - 85%, Rat - 86%.

Differentiated embryo-chondrocyte expressed gene 1 (DEC1) is an important protein involved in embryonic cell differentiation, proliferation, and apoptosis. It has also recently been shown to be regulated by hypoxia. The loss of DEC1 expression may be an early event in the development of lung cancer, while DEC1 may be up-regulated by hypoxia in other cancers and in more extreme hypoxia it may have a role in cell death.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BHLHE40
Applications ChIP Assay, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.10 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene BHLHE40
Gene Accession No. O14503
Gene Alias 1-Dec; basic helix-loop-helix domain containing, class B, 2; basic helix-loop-helix domain containing, class B2; basic helix-loop-helix family member e40; basic helix-loop-helix family, member e40; bHLHb2; Bhlhe40; C130042M06Rik; Class B basic helix-loop-helix protein 2; class E basic helix-loop-helix protein 40; Clast5; CR8; cytokine response gene 8; DEC1; differentially expressed in chondrocytes 1; differentially expressed in chondrocytes protein 1; differentiated embryo chondrocyte expressed gene 1; E47 interaction protein 1; EIP1; eip1 (E47 interaction protein 1); enhancer-of split and hairy-related protein 2; enhancer-of-split and hairy-related protein 2; FLJ99214; HLHB2; SHARP2; SHARP-2; stimulated by retinoic acid 14; stimulated by retinoic acid gene 13 protein; STRA13; Stra14
Gene Symbols BHLHE40
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human BHLHE40. Recombinant protein control fragment (Product #RP-102550).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8553
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.