missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BHLHE41 Monoclonal Antibody (OTI3H4), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA806354
This item is not returnable.
View return policy
Description
BHLHE41 may be a transcriptional repressor that represses both basal and activated transcription.Specifications
| BHLHE41 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9C0J9 | |
| BHLHE41 | |
| Human recombit protein fragment corresponding to amino acids 1-297 of human BHLHE41 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI3H4 | |
| Unconjugated | |
| BHLHE41 | |
| BHLHB3, DEC2, SHARP1, hDEC2 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 79365 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction