missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BHLHE41 Monoclonal Antibody (OTI8A8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA806509S
This item is not returnable.
View return policy
Description
BHLHE41 Monoclonal antibody specifically detects BHLHE41 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| BHLHE41 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9BRP8 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 84305 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI8A8 | |
| Unconjugated | |
| PYM1 | |
| 2-Dec; 6430520M22Rik; basic helix-loop-helix domain containing, class B, 3; basic helix-loop-helix domain containing, class B3; basic helix-loop-helix family member e41; basic helix-loop-helix family, member e41; bHLH transcriptional factor Dec2; Bhlhb2l; Bhlhb3; bHLHe41; Class B basic helix-loop-helix protein 3; class E basic helix-loop-helix protein 41; Dec2; differentially expressed in chondrocytes protein 2; Enhancer-of-split and hairy-related protein 1; hDEC2; mDEC2; SHARP1; SHARP-1 | |
| Full length human recombinant protein of human WIBG produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction