missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BHMT Monoclonal Antibody (OTI1E5), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA500961
This item is not returnable.
View return policy
Description
This gene encodes a cytosolic enzyme that catalyzes the conversion of betaine and homocysteine to dimethylglycine and methionine, respectively. Defects in this gene could lead to hyperhomocystinemia, but such a defect has not yet been observed.Specifications
| BHMT | |
| Monoclonal | |
| 0.7 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q93088 | |
| BHMT | |
| Full length human recombit protein of human BHMT produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| OTI1E5 | |
| Unconjugated | |
| BHMT | |
| BHMT1, HEL-S-61p | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 635 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction