missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ BI 6015

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Hepatocyte nuclear factor 4α (HNF4α) antagonist. Represses expression of known HNF4α target genes. Decreases HNF4α DNA binding. Exhibits cytotoxic activity in a range of tumor cell lines, including human hepatocellular carcinoma.
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 331.35 |
| Purity | >98% |
| Molecular Weight (g/mol) | 331.346 |
| Chemical Name or Material | BI 6015 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction