missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BI-847325, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
Description
BI-847325 is an ATP competitive dual inhibitor of MEK and aurora kinases (AK) with IC50 values of 4 and 15 nM for human MEK2 and AK-C, respectively.
Specifications
Specifications
| Chemical Name or Material | BI-847325 |
| Physical Form | Solid |
| CAS | 1207293-36-4 |
| Color | Yellow |
| Quantity | 25 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C29H28N4O2 |
| For Use With (Application) | Cancer-Kinase/protease |
| Solubility Information | DMSO : 16.67 mg/mL (35.88 mM; Need ultrasonic) |
| SMILES | O=C(NCC)C#CC1=CC(NC/2=O)=C(C=C1)C2=C(NC3=CC=C(CN(C)C)C=C3)/C4=CC=CC=C4 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction