Learn More
BIBR 1532, MedChemExpress
Product Code. 16497246
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
Product Code. 16497246
Brand: Medchem Express HY17353
MedChemExpress BIBR 1532 is a potent, selective and non-competitive telomerase inhibitor with IC50 of 100 nM in a cell-free assay.
BIBR 1532 is a potent, selective and non-competitive telomerase inhibitor with IC50 of 100 nM in a cell-free assay.
Specifications
Chemical Name or Material BIBR 1532
Physical Form Solid
CAS 321674-73-1
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C21H17NO3
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : ≥ 100 mg/mL (301.79 mM)
SMILES O=C(O)C1=CC=CC=C1NC(/C=C(C2=CC=C3C=CC=CC3=C2)\C)=O
Molecular Weight (g/mol) 331.36
Formula Weight 331.36
Percent Purity 99.22%
Grade Research
Show More Show Less
Research purposes only