missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BICD1 Antibody, PE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-78735PE
This item is not returnable.
View return policy
Description
BICD1 Polyclonal antibody specifically detects BICD1 in Human samples. It is validated for Western Blot, Immunoprecipitation, KnockDown
Specifications
| BICD1 | |
| Polyclonal | |
| Western Blot, Immunoprecipitation, Knockdown Validated | |
| Bicaudal D (Drosophila) homolog 1, bicaudal D homolog 1 (Drosophila), BICD, Bic-D 1, cytoskeleton-like bicaudal D protein homolog 1, protein bicaudal D homolog 1 | |
| The immunogen for this product maps to a region between residue 550 and 600 of human Bicaudal D Homolog 1 using the numbering given in entry NP_001705.2 (GeneID 636). | |
| 0.1 mL | |
| Neuroscience | |
| 636 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, Immunoprecipitation, KnockDown | |
| PE | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction