missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BID Antibody, Alexa Fluor™ 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB500-245AF350
This item is not returnable.
View return policy
Description
BID Polyclonal antibody specifically detects BID in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| BID | |
| Polyclonal | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| apoptic death agonist, BH3 interacting domain death agonist, BH3-interacting domain death agonist, BID isoform ES(1b), BID isoform L(2), BID isoform Si6, desmocollin type 4, FP497, Human BID coding sequence, MGC15319, MGC42355, p22 BID, tbid | |
| The immunogen this antibody was made to, maps to a region between residues 60 and 120 of human BH3 interacting domain death agonist using the number given in entry NP_932070.1 (GeneID 637). The epitope is found in isoforms 1 and 2 (but not isoform 3) of human BID. | |
| 0.1 mL | |
| Apoptosis, Cancer, Phospho Specific, Tumor Suppressors | |
| 637 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Alexa Fluor 350 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction