missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BID Antibody (OTI1G10), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70254APC
This item is not returnable.
View return policy
Description
BID Monoclonal antibody specifically detects BID in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
Specifications
| BID | |
| Monoclonal | |
| APC | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry, Immunocytochemistry | |
| OTI1G10 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence | |
| apoptic death agonist, BH3 interacting domain death agonist, BH3-interacting domain death agonist, BID isoform ES(1b), BID isoform L(2), BID isoform Si6, desmocollin type 4, FP497, Human BID coding sequence, MGC15319, MGC42355, p22 BID, tbid | |
| Full length human recombinant protein of human BID(NP_001187) produced in HEK293T cell. | |
| 0.1 mL | |
| Apoptosis, Cancer, Phospho Specific, Tumor Suppressors | |
| 637 | |
| Store at 4C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction