Learn More
Trajan Scientific and Medical Big Trap - Hydrocarbon
Click to view available options
Port Type:
1/4 in.
1/8 in.
Unit Size:
Each
Description
- High capacity filters ideal for bulk purification - capacity of 80 g organics, depending upon impurities
- Efficiency < 15 ppb
- Can be used where several instruments are plumbed from a single gas source
- Sintered stainless steel frits prevent particulate contamination
Specifications
Specifications
| For Use With (Application) | FID for air and hydrogen, PID when used with an oxygen filter |
| Model | Hydrocarbon |
| Volume (Metric) | 750 cm3 |
| Port Type | 1/8 in. |
| Description | Removes organic compounds from gas streams |
| For Use With | Big Trap Mounting Clip |
| Pressure | 250 psig |