Learn More
Trajan Scientific and Medical Big Trap - universal
For bulk purification applications or where several instruments are plumbed from a single source, a Big Trap gas filter is an ideal solution. A one-piece heavy walled alminium tube provides 750cm3 of capacity and a pressure rating of up to 250 psig
Brand:  Trajan Scientific and Medical 1035344
Additional Details : Weight : 2.00000kg
 View more versions of this product
Product Code. 15750979
  • € 333.00 / Each
Estimated Shipment: 17-05-2024
to see stock.
Add to basket
Description
Description
  • High capacity filters ideal for bulk purification - cumulative capacity level of 100μg/L of oxygen, moisture, hydrocarbons, carbon dioxide and carbon monoxide at a flow rate of 8L/min
  • Efficiency < 100 ppb
  • Can be used where several instruments are plumbed from a single gas source
  • Sintered stainless steel frits prevent particulate contamination
Specifications
Specifications
GC and GCMS applications where helium is employed
750 cm3
Removes oxygen, moisture, hydrocarbons, carbon dioxide and carbon monoxide
250 psig
Hydrocarbon
1/4 in.
Big Trap Mounting Clip
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
For research use only