Learn More
Trajan Scientific and Medical Big Trap - universal
For bulk purification applications or where several instruments are plumbed from a single source, a Big Trap gas filter is an ideal solution. A one-piece heavy walled alminium tube provides 750cm3 of capacity and a pressure rating of up to 250 psig
Brand: Trajan Scientific and Medical 1035344
Additional Details : Weight : 2.00000kg
Description
- High capacity filters ideal for bulk purification - cumulative capacity level of 100μg/L of oxygen, moisture, hydrocarbons, carbon dioxide and carbon monoxide at a flow rate of 8L/min
- Efficiency < 100 ppb
- Can be used where several instruments are plumbed from a single gas source
- Sintered stainless steel frits prevent particulate contamination
Specifications
GC and GCMS applications where helium is employed | |
750 cm3 | |
Removes oxygen, moisture, hydrocarbons, carbon dioxide and carbon monoxide | |
250 psig |
Hydrocarbon | |
1/4 in. | |
Big Trap Mounting Clip |
For research use only