missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ BIIE 0246 hydrochloride

Description
BIIE 0246 hydrochloride is a potent, selective and competitive nonpeptide antagonist for the neuropeptide Y Y2 receptor (IC50 = 15 nM). Displays > 650-fold selectivity for NPY Y2 over Y1, Y4 and Y5 receptors. Active in vivo.
Specifications
Specifications
| Chemical Name or Material | N-[(1 S)-4-[(Aminoiminomethyl)amino]-1-[[[2-(3,5-dioxo-1,2-diphenyl-1,2,4-triazolidin-4-yl)ethyl]amino]carbonyl]butyl]-1-[2-[4-(6,11-dihydro-6-oxo-5 H-dibenz[b,e]azepin-11-yl)-1-piperazinyl]-2-oxoethyl]-cyclopentaneacetamide hydrochloride |
| CAS | 2624332-04-1 |
| Quantity | 1 mg |
| Target | NPY Receptor Antagonists |
| Molecular Formula | C49H57N11O6.HCl |
| Purity | 0.98 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?