Learn More
Invitrogen™ BIK Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA599549
Product Code. 16683795
  • € 493.00 / 100µL
Description
Antibody detects endogenous levels of total BIK.
The Bik protein is known to interact with cellular and viral survival-promoting proteins, such as BCL2 and the Epstein-Barr virus in order to enhance programmed cell death. Because its activity is suppressed in the presence of survival-promoting proteins, this protein is suggested as a likely target for antiapoptotic proteins. This protein shares a critical BH3 domain with other death-promoting proteins, BAX and BAK.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
BIK
Polyclonal
Unconjugated
BIK
apoptosis inducer NBK; apoptosis-inducing NBK; BBC1; BCL2 interacting killer; bcl-2-interacting killer; BCL2-interacting killer; BCL2-interacting killer (apoptosis-inducing); bhikhari; BIK; Bik-like killer protein; Biklk; BIP 1; BIP1; Blk; BP 4; BP4; cb 60; natural born killer; NBK
Rabbit
Affinity chromatography
RUO
12124, 638
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
O70337, Q13323
BIK
A synthesized peptide derived from human BIK(Accession Q13323), corresponding to amino acid residues L7-L57.
100 μL
Primary
Human, Mouse, Monkey