missing translation for 'onlineSavingsMsg'
Learn More

Bikinin, MedChemExpress

Product Code. p-200096441
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16485906

Brand: Medchem Express HY12524

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Bikinin is a non-steroidal, ATP-competitive inhibitor of plant GSK-3/Shaggy-like kinases and activates BR (brassinosteroids) signaling.

Bikinin is a non-steroidal, ATP-competitive inhibitor of plant GSK-3/Shaggy-like kinases and activates BR (brassinosteroids) signaling.

Specifications

Chemical Name or Material Bikinin
CAS 188011-69-0
Color White
Purity Grade Notes Research
Molecular Formula C9H9BrN2O3
Quantity 100 mg
Synonym Abrasin
Solubility Information DMSO : ≥ 42 mg/mL (153.80 mM)
SMILES O=C(O)CCC(NC1=NC=C(Br)C=C1)=O
Molecular Weight (g/mol) 273.08
Formula Weight 273.08
Percent Purity 99.94%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.