missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bim Antibody (10B12), Alexa Fluor™ 532, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Bio-Techne NBP2-80103AF532
This item is not returnable.
View return policy
Description
Bim Monoclonal antibody specifically detects Bim in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence
Specifications
| Bim | |
| Monoclonal | |
| Alexa Fluor 532 | |
| BAM, bcl-2 interacting mediator of cell death, bcl-2 interacting protein Bim, Bcl2-interacting mediator of cell death, bcl2-L-11, BCL2-like 11 (apoptosis facilitator), bcl-2-like protein 11, bcl-2-related ovarian death agonist, BIM-alpha6, BIM-beta6, BIM-beta7, BIMBOD, BimEL, BimL | |
| Synthetic peptide corresponding to aa 20-40 of mouse BimL. | |
| 0.1 mL | |
| Apoptosis, Cancer, Immunology, Innate Immunity, Phospho Specific, Tumor Suppressors | |
| 10018 | |
| Store at 4°C in the dark. | |
| IgG2a κ |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence | |
| 10B12 | |
| 50mM Sodium Borate | |
| Rat | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Canine, Monkey | |
| Purified |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur