missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bim Antibody, Novus Biologicals™
Goat Polyclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NB100-1440
This item is not returnable.
View return policy
Description
Bim Polyclonal specifically detects Bim in Human samples. It is validated for Western Blot, Peptide ELISA.
Specifications
| Bim | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| BAM, bcl-2 interacting mediator of cell death, bcl-2 interacting protein Bim, Bcl2-interacting mediator of cell death, bcl2-L-11, BCL2-like 11 (apoptosis facilitator), bcl-2-like protein 11, bcl-2-related ovarian death agonist, BIM-alpha6, BIM-beta6, BIM-beta7, BIMBOD, BimEL, BimL | |
| Goat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| This antibody is expected to recognize at least reported isoforms Bim-alpha1 (BimABCD), Bim-alpha2 (BimACD) and Bim-alpha3 (BimAD). | |
| Store at -20C. Avoid freeze-thaw cycles. |
| Western Blot, ELISA | |
| 0.5 mg/mL | |
| Western Blot 0.5 - 2 ug/ml, Peptide ELISA Detection limit 1:16000 | |
| NP_619528 | |
| BCL2L11 | |
| Peptide with sequence C-FNAYYARRLEK corresponding to C-Terminus according to NP_619528.1, NP_619529.1, NP_996886.1. | |
| 0.1 mg | |
| Apoptosis, Cancer, Immunology, Innate Immunity, Tumor Suppressors | |
| 10018 | |
| Human | |
| IgG |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
For Research Use Only