Learn More
Macherey-Nagel™ BIO-LAB-TOP
Product Code. 10789531
Change view
Click to view available options
Dimensions (L x W):
48 x 60 cm
5000 x 48 cm
5000 x 60 cm
60 x 1000 cm
Quantity:
1 Ea.
100Pack
Unit Size:
Each
Pack of 100
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) Quantity Unit Size
10789531 48 x 60 cm 100Pack Pack of 100
10344461 5000 x 48 cm 1 Ea. Each
10542323 60 x 1000 cm 1 Ea. Each
10420053 5000 x 60 cm 1 Ea. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 10789531 Supplier Macherey-Nagel™ Supplier No. 740801
In Stock
Add to basket
Add to basket
Protection of laboratory surfaces from spills.
  • Filter paper, one side coated with polyethylene
Specifications
Product Type Surface Protection Paper
Dimensions (L x W) 48 x 60 cm
Thickness 0.22 mm
Quantity 100Pack