missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KGW-Isotherm BIO Liquid Nitrogen Container, Aluminum
Click to view available options
Capacity (Metric):
12.2 L
Unit Size:
Each
Description
- Aluminum container with compound materials in the neck range
- Low weight for easy handling
- Super-insolation foil in the vacuum space
- Low evaporation rate
- Six (6) canisters made of stainless steel
- Complies with international regulations regarding the transport of hazardous goods by road (ADR) or rail (RID)
- Includes: transport handle, vacuum valve, loose lied plug, canisters
Specifications
Specifications
| Capacity (Metric) | 12.2 L |
| Certifications/Compliance | ADR/RID |
| Color | White |
| For Use With (Application) | Storage and transport of biological materials |
| For Use With (Equipment) | Liquid nitrogen (LN2 |
| Includes | Handle, Vacuum valve, Plug, Canisters |
| Model | BIO |
| Static Evaporation Rate | 0.09 L/day |
| Static Holding Time | 130 Days |
| Height (Metric) Exterior | 630 mm |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction