Learn More
Gilac™ Biobased Dough Container
Biobased Dividing Dough Container is highly resistant to changes in temperature, highly flexible and shock-resistant.
Brand: Gilac™ G179491
Additional Details : Weight : 1.27000kg
Description
- Biobased dough storage container - 600 x 400.
- Stacking with and without lid, maximum stacking height capacity.
- Innovative design for even greater convenience:
- Rounded corners and bottom for easy cleaning.
- Vertical sides without inner edge avoids marking the dough. The entire surface of the container is usable.
- Completely flat bottom means dough detaches easily, no loss, no food deposit left.
- Optimised side reinforcements allows you to customise containers with your logo.
- Rounded outside corners guarantee a damage-free working environment.
- The dough container made from biosourced material is dye- and odour-free with a white appearance.
- Biobased material is made from natural renewable resources instead of limited energy sources such as fossil fuels.
- As a responsible French manufacturer, GILAC is now offering alternatives to standard materials, whilst maintaining an optimal level of quality and food safety in full compliance with current regulations.
- This dough container adds to the existing range of dough containers made with conventional materials.
Specifications
Dough Container | |
White | |
4 gal. |
Polyethylene | |
600 x 400 x 90 mm | |
15 L |