Learn More
Agilent BioTek BioCell Vessel and Adapter
Shop All Agilent BioTek Products
Click to view available options
Product Type:
Adapter Plate Assembly
Cuvette
For Use With (Application):
1cm absorbance measurements
1cm pathlength measurements
Unit Size:
Each
Description
BioCell Adapter Plate Assembly can hold up to 8 BioCell cuvets (BT7272051) for 1 cm pathlength measurements in a Agilent BioTek microplate spectrophotometer.
Specifications
Specifications
| Product Type | Adapter Plate Assembly |
| For Use With (Equipment) | Epoch, Synergy H1, Synergy HTX |
| For Use With (Application) | 1cm pathlength measurements |